COM448628120 Combi-Blocks, Inc.

(4-Cyclohexyl-phenyl)-(4-isopropyl-phenyl)-amine

  • MDL number MFCD07784078
  • Cas Number : 886365-92-0
  • Molecular Weight : 293.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8703

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8703
Availability details for
Quantity
  • Availability
  • Add to Cart