COM448628123 Combi-Blocks, Inc.

(4'-Formyl-biphenyl-4-yl)-acetic acid

  • MDL number MFCD03426487
  • Cas Number : 669713-90-0
  • Molecular Weight : 240.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8708

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8708
Availability details for
Quantity
  • Availability
  • Add to Cart