COM448628126 Combi-Blocks, Inc.

(4'-Methoxy-biphenyl-2-yl)-acetic acid

  • MDL number MFCD03426470
  • Cas Number : 108478-21-3
  • Molecular Weight : 242.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8712

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8712
Availability details for
Quantity
  • Availability
  • Add to Cart