COM448628128 Combi-Blocks, Inc.

(4-Methyloxolan-3-yl)methanol

  • MDL number MFCD17926174
  • Cas Number : 22600-87-9
  • Molecular Weight : 116.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8714

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8714
Availability details for
Quantity
  • Availability
  • Add to Cart