COM448628133 Combi-Blocks, Inc.

(4-Oxo-cyclohexylmethyl)-carbamic acid benzyl ester

  • MDL number MFCD11501248
  • Cas Number : 869895-17-0
  • Molecular Weight : 261.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8720

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8720
Availability details for
Quantity
  • Availability
  • Add to Cart