COM448628135 Combi-Blocks, Inc.

(4-Oxo-piperidin-1-yl)-phenyl-acetic acid

  • MDL number MFCD07369978
  • Cas Number : 886363-69-5
  • Molecular Weight : 233.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8722

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8722
Availability details for
Quantity
  • Availability
  • Add to Cart