COM448628145 Combi-Blocks, Inc.

(4S)-4-(4-Trifluoromethyl-benzyl)-l-glutamic acid

  • MDL number MFCD07784142
  • Cas Number : 207508-60-9
  • Molecular Weight : 305.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8740

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8740
Availability details for
Quantity
  • Availability
  • Add to Cart