COM448628148 Combi-Blocks, Inc.

(5,6,7,8-Tetrahydro-[1,8]naphthyridin-2-yl)-acetic acid methyl ester 2hcl

  • MDL number MFCD18711605
  • Cas Number : 1159823-05-8
  • Molecular Weight : 279.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8747

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8747
Availability details for
Quantity
  • Availability
  • Add to Cart