COM448628153 Combi-Blocks, Inc.

(5S)-5-(Hydroxymethyl)-3,3-dimethyl-2-pyrrolidinone

  • MDL number MFCD18711408
  • Cas Number : 156088-46-9
  • Molecular Weight : 143.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8760

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8760
Availability details for
Quantity
  • Availability
  • Add to Cart