COM448628161 Combi-Blocks, Inc.

(6S)-2-Amino-6-propionamidotetrahydrobenzothiazole

  • MDL number MFCD07369803
  • Cas Number : 106006-84-2
  • Molecular Weight : 225.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8772

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8772
Availability details for
Quantity
  • Availability
  • Add to Cart