COM448628162 Combi-Blocks, Inc.

(8-Boc-5,6,7,8-tetrahydro-[1,8]naphthyridin-2-yl)-acetic acid methyl ester

  • MDL number MFCD09025326
  • Cas Number : 925889-81-2
  • Molecular Weight : 306.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8774

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8774
Availability details for
Quantity
  • Availability
  • Add to Cart