COM448628170 Combi-Blocks, Inc.

(E)-2-(4-Fluorobenzylideneamino)propane-1,3-diol

  • MDL number MFCD09788594
  • Cas Number : 951623-97-5
  • Molecular Weight : 197.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8790

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8790
Availability details for
Quantity
  • Availability
  • Add to Cart