COM448628180 Combi-Blocks, Inc.

(E)-4-(4-Aminostyryl)benzenamine

  • MDL number MFCD00044493
  • Cas Number : 54760-75-7
  • Molecular Weight : 210.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8805

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8805
Availability details for
Quantity
  • Availability
  • Add to Cart