COM448628181 Combi-Blocks, Inc.

(E)-Ethyl 3-cyano-4-((dimethylamino) methyleneamino)phenylcarbamate

  • MDL number MFCD11040517
  • Cas Number : 1033194-59-0
  • Molecular Weight : 260.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8807

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8807
Availability details for
Quantity
  • Availability
  • Add to Cart