COM448628183 Combi-Blocks, Inc.

(E)-N'-(4-Amino-2-cyanophenyl)-n,n-dimethylformimidamide

  • MDL number MFCD09998252
  • Cas Number : 194423-17-1
  • Molecular Weight : 188.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8809

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8809
Availability details for
Quantity
  • Availability
  • Add to Cart