COM448628186 Combi-Blocks, Inc.

(Pyrrol-3-yl)-acetic acid

  • MDL number MFCD09800534
  • Cas Number : 86688-96-2
  • Molecular Weight : 125.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1H-pyrrol-3-ylacetic acid
Inchi Code 1S/C6H7NO2/c8-6(9)3-5-1-2-7-4-5/h1-2,4,7H,3H2,(H,8,9)
Inchi Key LAZKLUQBPMKCFN-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8815

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8815
Availability details for
Quantity
  • Availability
  • Add to Cart