COM448628206 Combi-Blocks, Inc.

(R)-1-(3,4-Dimethoxy-benzyl)-6,7-dimethoxy-1,2,3,4-tetrahydro-isoquinoline, HCl

  • MDL number MFCD03788189
  • Cas Number : 54417-53-7
  • Molecular Weight : 379.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8856

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8856
Availability details for
Quantity
  • Availability
  • Add to Cart