COM448628207 Combi-Blocks, Inc.

(R)-1-(4-Isopropyl-2-thioxothiazolidin-3-yl)ethanone

  • MDL number MFCD10570251
  • Cas Number : 121929-87-1
  • Molecular Weight : 203.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8859

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8859
Availability details for
Quantity
  • Availability
  • Add to Cart