COM448628211 Combi-Blocks, Inc.

(R)-1,2,3,4-Tetrahydro-2-naphthylamine

  • MDL number MFCD00671631
  • Cas Number : 21966-60-9
  • Molecular Weight : 147.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8872

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8872
Availability details for
Quantity
  • Availability
  • Add to Cart