COM448628214 Combi-Blocks, Inc.

(R)-1,4-Benzodioxan-2-carboxypiperazine

  • MDL number MFCD06796578
  • Cas Number : 860173-98-4
  • Molecular Weight : 248.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8876

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8876
Availability details for
Quantity
  • Availability
  • Add to Cart