COM448628220 Combi-Blocks, Inc.

(R)-1-Benzyl-6-hydroxymethyl-piperazin-2-one

  • MDL number MFCD06796561
  • Cas Number : 1217657-81-2
  • Molecular Weight : 220.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8887

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8887
Availability details for
Quantity
  • Availability
  • Add to Cart