COM448628233 Combi-Blocks, Inc.

(R)-1-N-Boc-pipecolamide

  • MDL number MFCD03840195
  • Cas Number : 848488-91-5
  • Molecular Weight : 228.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8910

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8910
Availability details for
Quantity
  • Availability
  • Add to Cart