COM448628235 Combi-Blocks, Inc.

(R)-1-N-Cbz-2-chloromethyl-pyrrolidine

  • MDL number MFCD08061231
  • Cas Number : 39945-49-8
  • Molecular Weight : 253.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8913

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8913
Availability details for
Quantity
  • Availability
  • Add to Cart