COM448628245 Combi-Blocks, Inc.

(R)-2-(Piperidin-3-yl)acetic acid hcl

  • MDL number MFCD20261469
  • Cas Number : 1334509-89-5
  • Molecular Weight : 179.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8931
Availability details for
Quantity
  • Availability
  • Add to Cart