COM448628257 Combi-Blocks, Inc.

(R)-2-Cbz-amino-butane-1,4-diol

  • MDL number MFCD03426374
  • Cas Number : 672309-94-3
  • Molecular Weight : 239.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8963

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8963
Availability details for
Quantity
  • Availability
  • Add to Cart