COM448628260 Combi-Blocks, Inc.

(R)-2-Methylpiperazine 2hcl

  • MDL number MFCD11053491
  • Cas Number : 75336-89-9
  • Molecular Weight : 173.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8971

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8971
Availability details for
Quantity
  • Availability
  • Add to Cart