COM448628274 Combi-Blocks, Inc.

(R)-3-(Benzyloxycarbonylamino)-5-methylhexanoic acid

  • MDL number MFCD06223035
  • Cas Number : 118247-77-1
  • Molecular Weight : 279.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8993

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8993
Availability details for
Quantity
  • Availability
  • Add to Cart