COM448628288 Combi-Blocks, Inc.

(R)-3-Amino-4,4-dimethyl-pentanoic acid methyl ester, HCl

  • MDL number MFCD12068631
  • Cas Number : 1422051-73-7
  • Molecular Weight : 195.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9025

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9025
Availability details for
Quantity
  • Availability
  • Add to Cart