COM448628290 Combi-Blocks, Inc.

(R)-3-Amino-4-methyl-pentanoic acid methyl ester, HCl

  • MDL number MFCD12068619
  • Cas Number : 172823-13-1
  • Molecular Weight : 181.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9027

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9027
Availability details for
Quantity
  • Availability
  • Add to Cart