COM448628293 Combi-Blocks, Inc.

(R)-3-Amino-dihydro-furan-2,5-dione, HCl

  • MDL number MFCD09878755
  • Cas Number : 143394-93-8
  • Molecular Weight : 151.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9031

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9031
Availability details for
Quantity
  • Availability
  • Add to Cart