COM448628299 Combi-Blocks, Inc.

(R)-3-Boc-4-(methoxymethylcarbamoyl)-2,2-dimethyloxazolidine

  • MDL number MFCD09878778
  • Cas Number : 167102-62-7
  • Molecular Weight : 288.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9039

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9039
Availability details for
Quantity
  • Availability
  • Add to Cart