COM448628302 Combi-Blocks, Inc.

(R)-3-Chloro-1-(3,4-dichloro-phenyl)-propan-1-ol

  • MDL number MFCD07784068
  • Cas Number : 1233501-44-4
  • Molecular Weight : 239.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9042

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9042
Availability details for
Quantity
  • Availability
  • Add to Cart