COM448628303 Combi-Blocks, Inc.

(R)-3-Methyl-pentanoic acid

  • MDL number MFCD06809587
  • Cas Number : 16958-25-1
  • Molecular Weight : 116.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9044

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9044
Availability details for
Quantity
  • Availability
  • Add to Cart