COM448628308 Combi-Blocks, Inc.

(R)-3-N-Cbz-aminovaleric acid

  • MDL number MFCD11053569
  • Molecular Weight : 251.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9050

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9050
Availability details for
Quantity
  • Availability
  • Add to Cart