COM448628311 Combi-Blocks, Inc.

(R)-3-Piperidine acetate ethyl ester hcl

  • MDL number MFCD09955432
  • Cas Number : 1233200-48-0
  • Molecular Weight : 207.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9054

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9054
Availability details for
Quantity
  • Availability
  • Add to Cart