COM448628312 Combi-Blocks, Inc.

(R)-3-Pyrrolidineacetic acid hcl

  • MDL number MFCD06858513
  • Cas Number : 122442-01-7
  • Molecular Weight : 165.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9055

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9055
Availability details for
Quantity
  • Availability
  • Add to Cart