COM448628314 Combi-Blocks, Inc.

(R)-4-(4-Aminomethyl-benzyl)-oxazolidin-2-one

  • MDL number MFCD08061184
  • Cas Number : 1217732-05-2
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9058

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9058
Availability details for
Quantity
  • Availability
  • Add to Cart