COM448628320 Combi-Blocks, Inc.

(R)-4-Benzyl-1,3-thiazolidine-2-one

  • MDL number MFCD06796568
  • Cas Number : 1217647-47-6
  • Molecular Weight : 193.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9064

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9064
Availability details for
Quantity
  • Availability
  • Add to Cart