COM448628323 Combi-Blocks, Inc.

(R)-4-Boc-1-cbz-2-hydroxymethylpiperazine

  • MDL number MFCD09261339
  • Cas Number : 930782-89-1
  • Molecular Weight : 350.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 97

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9068
Availability details for
Quantity
  • Availability
  • Add to Cart