COM448628328 Combi-Blocks, Inc.

(R)-4-Cyanothiazolidine, HCl

  • MDL number MFCD02683061
  • Cas Number : 391248-17-2
  • Molecular Weight : 150.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9076

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9076
Availability details for
Quantity
  • Availability
  • Add to Cart