COM448628329 Combi-Blocks, Inc.

(R)-4-Cyano-thiazolidine-3-carboxylic acid tert-butyl ester

  • MDL number MFCD02683054
  • Cas Number : 391248-15-0
  • Molecular Weight : 214.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9077

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9077
Availability details for
Quantity
  • Availability
  • Add to Cart