COM448628339 Combi-Blocks, Inc.

(R)-5-N-Boc-amino-3-hydroxyl-pentanioc acid

  • MDL number MFCD03839831
  • Cas Number : 1217830-91-5
  • Molecular Weight : 233.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9095

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9095
Availability details for
Quantity
  • Availability
  • Add to Cart