COM448628341 Combi-Blocks, Inc.

(R)-6-Hydroxymethyl-1-(4-hydroxy-phenyl)-piperazin-2-one

  • MDL number MFCD06796629
  • Cas Number : 1217608-77-9
  • Molecular Weight : 222.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9100

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9100
Availability details for
Quantity
  • Availability
  • Add to Cart