COM448628348 Combi-Blocks, Inc.

(R)-Cyclohexylhydroxyphenylacetic acid

  • MDL number MFCD09753427
  • Cas Number : 20585-39-1
  • Molecular Weight : 234.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9111

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9111
Availability details for
Quantity
  • Availability
  • Add to Cart