COM448628360 Combi-Blocks, Inc.

(R)-N-(2-Amino-4,5,6,7-tetrahydro-benzothiazol-6-yl)-propionamide

  • MDL number MFCD07369802
  • Cas Number : 106006-85-3
  • Molecular Weight : 225.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9137

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9137
Availability details for
Quantity
  • Availability
  • Add to Cart