COM448628361 Combi-Blocks, Inc.

(R)-N-(2-Hydroxy-2-methyl-1-phenyl-propyl)-4-methyl-benzenesulfonamide

  • MDL number MFCD09025333
  • Cas Number : 620627-46-5
  • Molecular Weight : 319.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9138

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9138
Availability details for
Quantity
  • Availability
  • Add to Cart