COM448628371 Combi-Blocks, Inc.

(R)-Phenyl-p-tolylsulfoxid

  • MDL number MFCD11707196
  • Cas Number : 16491-20-6
  • Molecular Weight : 216.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9153

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9153
Availability details for
Quantity
  • Availability
  • Add to Cart