COM448628389 Combi-Blocks, Inc.

(S)-(-)-Methylsuccinic acid dimethyl ester

  • MDL number MFCD00134572
  • Cas Number : 63163-08-6
  • Molecular Weight : 160.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9182

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9182
Availability details for
Quantity
  • Availability
  • Add to Cart