COM448628395 Combi-Blocks, Inc.

(S)-(1-Benzyl-pyrrolidin-3-yl)-methanol

  • MDL number MFCD09608006
  • Cas Number : 78914-69-9
  • Molecular Weight : 191.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9193

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9193
Availability details for
Quantity
  • Availability
  • Add to Cart