COM448628396 Combi-Blocks, Inc.

(S)-(1-Hydroxymethyl-2-pyrrolidin-1-yl-ethyl)-methyl-carbamic acid benzyl ester

  • MDL number MFCD03426459
  • Cas Number : 675602-76-3
  • Molecular Weight : 292.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9195

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9195
Availability details for
Quantity
  • Availability
  • Add to Cart